PRIH93ED8092 Princeton BioMolecular Research, Inc.

(1Z)-2-methylpropanal thiosemicarbazone

  • MDL number MFCD30744951
  • Molecular Weight : 145.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart