PRIH93ED8097 Princeton BioMolecular Research, Inc.

(2E)-N-ethyl-2-(phenylhydrazono)propanamide

  • MDL number MFCD30744957
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart