PRIH93ED809B Princeton BioMolecular Research, Inc.

5-Bromo-2-(2-p-tolyloxy-ethylsulfanyl)-1H-benzoimidazole

  • MDL number MFCD05838209
  • Molecular Weight : 363.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart