PRIH93ED809F Princeton BioMolecular Research, Inc.

8-[2-(4-Methoxy-phenoxy)-ethylsulfanyl]-9H-purin-6-ylamine

  • MDL number MFCD02197983
  • Molecular Weight : 317.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart