PRIH93ED80A0 Princeton BioMolecular Research, Inc.

9-Ethyl-3-[2-(4-methoxy-phenoxy)-ethylsulfanyl]-9H-benzo[4,5]imidazo[2,1-c][1,2,4]triazole

  • MDL number MFCD06755497
  • Molecular Weight : 368.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart