PRIH93ED80AA Princeton BioMolecular Research, Inc.

6-[(2-methyl-1H-indol-3-yl)methyl]-3,5-dihydro-1H-imidazo[4,5-f]benzimidazol-2-one

  • MDL number MFCD31418282
  • Molecular Weight : 317.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart