PRIH93ED80D1 Princeton BioMolecular Research, Inc.

3-(2-Hydroxy-1,1-dimethyl-ethyl)-1H-quinoxalin-2-one

  • MDL number MFCD24616297
  • Cas Number : 1239897-92-7
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart