PRIH93ED80E2 Princeton BioMolecular Research, Inc.

2-(dodecylthio)[1,2,4]triazolo[1,5-a]pyrimidine-6-carbaldehyde

  • MDL number MFCD31727636
  • Molecular Weight : 348.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart