PRIH93ED80E6 Princeton BioMolecular Research, Inc.

{4-[(7-hydroxy-4-methyl-2-oxo-2H-chromen-8-yl)methyl]piperazin-1-yl}acetic acid

  • MDL number MFCD31727643
  • Molecular Weight : 332.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart