PRIH93ED80F3 Princeton BioMolecular Research, Inc.

2-[1-(4-fluorophenyl)-1H-tetrazol-5-yl]-4,6-dimethylthieno[2,3-b]pyridin-3-amine

  • MDL number MFCD24539914
  • Molecular Weight : 340.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart