PRIH93ED8106 Princeton BioMolecular Research, Inc.

{[2-(3,5-dimethylpiperidin-1-yl)-2-oxoethyl]thio}acetic acid

  • MDL number MFCD03719776
  • Molecular Weight : 245.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart