PRIH93ED8109 Princeton BioMolecular Research, Inc.

3-(2,4-dinitrophenyl)-3-methylbutanoic acid

  • MDL number MFCD31944596
  • Molecular Weight : 268.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart