PRIH93ED811C Princeton BioMolecular Research, Inc.

4-ethyl-5-methoxy-2-[5-methyl-4-(4-methyl-1,3-thiazol-2-yl)-1H-pyrazol-3-yl]phenol

  • MDL number MFCD01792673
  • Molecular Weight : 329.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart