PRIH93ED8124 Princeton BioMolecular Research, Inc.

6-ethyl-2-methyl-3-(4-methyl-1,3-thiazol-2-yl)-7-[(4-nitrobenzyl)oxy]-4H-chromen-4-one

  • MDL number MFCD02673933
  • Molecular Weight : 436.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart