PRIH93ED8131 Princeton BioMolecular Research, Inc.

2-benzylquinazolin-4(1H)-one

  • MDL number MFCD06650414
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart