PRIH93ED8137 Princeton BioMolecular Research, Inc.

2-ethyl-6-isonicotinoyl-5,11-dihydro-4H-thieno[3',2':5,6]pyrimido[1,2-b]isoquinolin-4-one

  • MDL number MFCD07423383
  • Molecular Weight : 387.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart