PRIH93ED8147 Princeton BioMolecular Research, Inc.

5-bromo-6-methoxy-8-[(E)-2-nitrovinyl]chromane

  • MDL number MFCD01910693
  • Molecular Weight : 314.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart