PRIH93ED8153 Princeton BioMolecular Research, Inc.

7-methoxy-2-methyl-3-(1,3-thiazol-4-yl)-4H-chromene-4-thione

  • MDL number MFCD01916972
  • Molecular Weight : 289.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart