PRIH93ED815A Princeton BioMolecular Research, Inc.

(2E)-1-(1-hydroxy-2-naphthyl)-3-(4-methoxyphenyl)prop-2-en-1-one

  • MDL number MFCD00855686
  • Molecular Weight : 304.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart