PRIH93ED8160 Princeton BioMolecular Research, Inc.

methyl {[5-methyl-3-(4-methyl-1,3-thiazol-2-yl)-4-oxo-4H-chromen-7-yl]oxy}acetate

  • MDL number MFCD01544798
  • Molecular Weight : 345.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart