PRIH93ED8168 Princeton BioMolecular Research, Inc.

2-amino-N'-[(1Z)-3-amino-1H-isoindol-1-ylidene]benzohydrazide

  • MDL number MFCD03480272
  • Molecular Weight : 279.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart