PRIH93ED8172 Princeton BioMolecular Research, Inc.

N-(5-methoxy-3-methyl-2,3-dihydro-1-benzofuran-6-yl)-2-morpholin-4-ylacetamide

  • MDL number MFCD07423384
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart