PRIH93ED8184 Princeton BioMolecular Research, Inc.

1,3-dichloroisoindolo[2,1-a]quinazolin-5(11H)-one

  • MDL number MFCD02691156
  • Molecular Weight : 303.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart