PRIH93ED8189 Princeton BioMolecular Research, Inc.

(12E)-12-(pyridin-4-ylmethylene)-8,9-dihydro-7H-cyclopenta[4',5']thieno[3',2':5,6]pyrimido[2,1-a]isoindol-6(12H)-one

  • MDL number MFCD06650422
  • Molecular Weight : 369.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart