PRIH93ED8191 Princeton BioMolecular Research, Inc.

(1E)-2-[4-(6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)phenyl]isoindolin-1-imine

  • MDL number MFCD02720463
  • Molecular Weight : 343.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart