PRIH93ED819B Princeton BioMolecular Research, Inc.

ethyl 3-{[3-(4-oxo-1,2,3-benzotriazin-3(4H)-yl)propanoyl]amino}benzoate

  • MDL number MFCD06650457
  • Molecular Weight : 366.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart