PRIH93ED819F Princeton BioMolecular Research, Inc.

6-({[(3,4,7-trimethyl-2-oxo-2H-chromen-5-yl)oxy]acetyl}amino)hexanoic acid

  • MDL number MFCD03851080
  • Molecular Weight : 375.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart