PRIH93ED81A9 Princeton BioMolecular Research, Inc.

N-(4-bromo-2-fluorophenyl)-6-(4-oxo-1,2,3-benzotriazin-3(4H)-yl)hexanamide

  • MDL number MFCD06650476
  • Molecular Weight : 433.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart