PRIH93ED81B1 Princeton BioMolecular Research, Inc.

3-{1-[(4a-hydroxy-1-phenyloctahydroisoquinolin-2(1H)-yl)carbonyl]propoxy}-7,8,9,10-tetrahydro-6H-benzo[c]chromen-6-one

  • MDL number MFCD06650484
  • Molecular Weight : 515.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart