PRIH93ED81BE Princeton BioMolecular Research, Inc.

13-(5-bromo-2-furoyl)-5-ethyl[1]benzofuro[2',3':5,6]pyrimido[2,1-a]isoindol-6(5H)-one

  • MDL number MFCD06650495
  • Molecular Weight : 475.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart