PRIH93ED81C5 Princeton BioMolecular Research, Inc.

N,N'-bis[4-(trifluoromethoxy)phenyl]urea

  • MDL number MFCD00277528
  • Molecular Weight : 380.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart