PRIH93ED81D0 Princeton BioMolecular Research, Inc.

2-{2-ethyl-5-hydroxy-4-[4-(4-methyl-1,3-thiazol-2-yl)-1H-pyrazol-3-yl]phenoxy}-N-(2-hydroxyethyl)propanamide

  • MDL number MFCD01089081
  • Molecular Weight : 416.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart