PRIH93ED81DB Princeton BioMolecular Research, Inc.

5-[(E)-(2-methyl-1,3-benzothiazol-6-yl)diazenyl]-4-phenyl-1,3-thiazol-2-amine

  • MDL number MFCD06650537
  • Molecular Weight : 351.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart