PRIH93ED81E5 Princeton BioMolecular Research, Inc.

2-(2-methyl-3-furyl)-5a,9a-dihydro-5H-[1,2,4]triazolo[5,1-a]isoindole

  • MDL number MFCD08836204
  • Molecular Weight : 239.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart