PRIH93ED81F1 Princeton BioMolecular Research, Inc.

6-amino-3-methyl-5-(1-naphthyl)-1-phenylpyridazin-4(1H)-one

  • MDL number MFCD08836597
  • Molecular Weight : 327.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart