PRIH93ED8203 Princeton BioMolecular Research, Inc.

6-amino-2-(4-nitrophenyl)-1H-benzo[de]isoquinoline-1,3(2H)-dione

  • MDL number MFCD09983183
  • Molecular Weight : 333.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart