PRIH93ED820C Princeton BioMolecular Research, Inc.

2-ethylthieno[3',2':5,6]pyrimido[2,1-a]isoindol-4(10H)-one

  • MDL number MFCD09983185
  • Molecular Weight : 268.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart