PRIH93ED821F Princeton BioMolecular Research, Inc.

6-fluoro-2',5'-dioxo-2,3-dihydrospiro[chromene-4,4'-imidazolidine]-2-carboxamide

  • MDL number MFCD00873153
  • Molecular Weight : 279.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart