PRIH93ED8230 Princeton BioMolecular Research, Inc.

4-amino-5-chloro-2-methylpyridazin-3(2H)-one

  • MDL number MFCD20690929
  • Molecular Weight : 159.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart