PRIH93ED8241 Princeton BioMolecular Research, Inc.

4-(methoxycarbonyl)phenyl 2,3-diphenylquinoxaline-6-carboxylate

  • MDL number MFCD03022306
  • Molecular Weight : 460.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart