PRIH93ED8249 Princeton BioMolecular Research, Inc.

1,7,7-trimethyl-4-(morpholin-4-ylcarbonyl)bicyclo[2.2.1]heptan-2-one

  • MDL number MFCD02641192
  • Molecular Weight : 265.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart