PRIH93ED824B Princeton BioMolecular Research, Inc.

4,7,7-trimethyl-N-(3-morpholin-4-ylpropyl)-3-oxobicyclo[2.2.1]heptane-1-carboxamide

  • MDL number MFCD03659950
  • Molecular Weight : 322.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart