PRIH93ED825A Princeton BioMolecular Research, Inc.

4,7,8,11,11-pentamethyl-3,4-dihydro-1,4-methanophenazine-1(2H)-carboxylic acid

  • MDL number MFCD03850458
  • Molecular Weight : 310.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart