PRIH93ED826B Princeton BioMolecular Research, Inc.

7,7-dimethyl-2,3-dioxo-N-[2-(trifluoromethyl)phenyl]bicyclo[2.2.1]heptane-1-carboxamide

  • MDL number MFCD06016637
  • Molecular Weight : 339.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart