PRIH93ED826E Princeton BioMolecular Research, Inc.

6-pyridin-2-ylhexanoic acid

  • MDL number MFCD06496347
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart