PRIH93ED8277 Princeton BioMolecular Research, Inc.

2-(1,3-benzothiazol-2-yl)nicotinohydrazide

  • MDL number MFCD06742783
  • Molecular Weight : 270.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart