PRIH93ED8282 Princeton BioMolecular Research, Inc.

5-chloro-N-methyl-N-(1-methylpiperidin-4-yl)-2-(methylthio)pyrimidine-4-carboxamide

  • MDL number MFCD08836420
  • Molecular Weight : 314.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart