PRIH93ED8293 Princeton BioMolecular Research, Inc.

1-(3,4-dimethylphenyl)-2,6,6-trimethyl-1,5,6,7-tetrahydro-4H-indol-4-one

  • MDL number MFCD05987778
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart