PRIH93ED829C Princeton BioMolecular Research, Inc.

3-methyl-4-oxo-4,7-dihydro-5H-thiopyrano[3,4-b]furan-2-carboxylic acid

  • MDL number MFCD06001747
  • Molecular Weight : 212.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart