PRIH93ED82C4 Princeton BioMolecular Research, Inc.

(1Z)-2-pyridin-2-ylcyclopentanone oxime

  • MDL number MFCD03665658
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart